2-Bromo-6-fluoropyridin-3-ol ≥97% research grade

Supplier: AOBChem USA

Warning

29481-25G 29481-5G
76901-268EA 300.24 USD
76901-268 76901-270
2-Bromo-6-fluoropyridin-3-ol ≥97% research grade
2-Bromo-6-fluoropyridin-3-ol
Formula: C₅H₃BrFNO
Storage Temperature: Ambient
MDL Number: MFCD18257625
CAS Number: 1256822-94-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR