2-Chloro-1,4-difluoro-3-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

28931-1G 28931-25G 28931-5G
76901-084EA 106.09 USD
76901-084 76901-086 76901-088
2-Chloro-1,4-difluoro-3-methylbenzene ≥97%
2-Chloro-1,4-difluoro-3-methylbenzene
Formula: C₇H₅ClF₂
MW: 162.56 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12922611
CAS Number: 90292-64-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR