2-Chloro-6-(trifluoromethyl)benzyl alcohol ≥97%

Supplier: AOBChem USA

Warning

28405-1G 28405-25G 28405-5G
76900-984EA 71.09 USD
76900-984 76900-986 76900-988
2-Chloro-6-(trifluoromethyl)benzyl alcohol ≥97%
2-Chloro-6-(trifluoromethyl)benzyl alcohol

Precaución: for Research Use Only

Formula: C₈H₆ClF₃O
Storage Temperature: Refrigerator
MDL Number: MFCD00049315
CAS Number: 886500-21-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR