2-Fluoro-4-methylbenzeneboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

27304-25G 27304-5G
76900-654EA 516.66 USD
76900-654 76900-656
2-Fluoro-4-methylbenzeneboronic acid pinacol ester ≥97%
2-Fluoro-4-methylbenzeneboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₈BFO₂
Storage Temperature: Ambient
MDL Number: MFCD18779806
CAS Number: 1165936-03-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR