2,4-Difluoro-3-iodophenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

24843-25G 24843-5G
76899-666EA 453.01 USD
76899-666 76899-668
2,4-Difluoro-3-iodophenylboronic acid ≥97%
2,4-Difluoro-3-iodophenylboronic acid
Formula: C₆H₄BF₂IO₂
Storage Temperature: Ambient
MDL Number: MFCD22628229
CAS Number: 1451393-12-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR