3-Bromo-4-iodophenol ≥97%

Supplier: AOBChem USA

Warning

56807-1G 56807-25G 56807-500MG 56807-5G
76917-284EA 206.88 USD
76917-284 76917-286 76917-288 76917-290
3-Bromo-4-iodophenol ≥97%
3-Bromo-4-iodophenol

Precaución: for Research Use Only

Formula: C₆H₄BrIO
Storage Temperature: Refrigerator
MDL Number: MFCD17000017
CAS Number: 1037298-05-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR