4-Isoquinolineboronic acid pinacol ester ≥97%
Supplier: AOBChem USA
|
Warning
|
Synonyms:
4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)isoquinoline
53400-25G
53400-5G
76917-020EA
573.95
USD
76917-020
76917-022
4-Isoquinolineboronic acid pinacol ester ≥97%
4-Isoquinolineboronic acid pinacol ester
Precaución: for Research Use Only
Formula:
C₁₅H₁₈BNO₂ MW: 255.1199 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD05663891 CAS Number: 685103-98-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...