3-Formyl-2-methoxyphenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Methoxy-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde

32959-1G 32959-25G 32959-500MG 32959-5G
76914-536EA 126.25 USD
76914-536 76914-538 76914-540 76914-542
3-Formyl-2-methoxyphenylboronic acid pinacol ester ≥97%
3-Formyl-2-methoxyphenylboronic acid pinacol ester
Formula: C₁₄H₁₉BO₄
MW: 262.109 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22495020
CAS Number: 1356638-77-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR