3-Methoxy-4-(trifluoromethoxy)phenol ≥97%

Supplier: AOBChem USA

Warning

32320-1G 32320-25G 32320-500MG 32320-5G
76914-208EA 379.81 USD
76914-208 76914-210 76914-212 76914-214
3-Methoxy-4-(trifluoromethoxy)phenol ≥97%
3-Methoxy-4-(trifluoromethoxy)phenol
Formula: C₈H₇F₃O₃
MW: 208.135 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18397425
CAS Number: 1261571-61-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR