4-Bromo-2-chloro-5-fluoro-1-iodobenzene ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-Bromo-2-chloro-5-fluoroiodobenzene, 1-Bromo-5-chloro-2-fluoro-4-iodobenzene

26395-1G 26395-25G 26395-5G
76912-370EA 88.06 USD
76912-370 76912-372 76912-374
4-Bromo-2-chloro-5-fluoro-1-iodobenzene ≥97%
4-Bromo-2-chloro-5-fluoro-1-iodobenzene

Precaución: for Research Use Only

Formula: C₆H₂BrClFI
MW: 335.34 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09878157
CAS Number: 1000572-73-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR