Bis(4-formylphenyl) ether ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-(4-Formylphenoxy)benzaldehyde, 4,4'-Diformyldiphenyl ether, 4,4'-Oxydibenzaldehyde

24645-1G 24645-25G 24645-5G
76911-748EA 62.6 USD
76911-748 76911-750 76911-752
Bis(4-formylphenyl) ether ≥97%
Bis(4-formylphenyl) ether

Precaución: for Research Use Only

Formula: C₁₄H₁₀O₃
MW: 226.23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08276812
CAS Number: 2215-76-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR