4-Bromo-N,N-dimethylpyridin-2-amine ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-Bromo-2-(dimethylamino)pyridine

19247-1G 19247-25G 19247-5G
76909-626EA 109.28 USD
76909-626 76909-628 76909-630
4-Bromo-N,N-dimethylpyridin-2-amine ≥97%
4-Bromo-N,N-dimethylpyridin-2-amine
Formula: C₇H₉BrN₂
MW: 201.0638 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11869615
CAS Number: 946000-27-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR