3'-Methyl-2,2,2,4'-tetrafluoroacetophenone ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2,2,2-Trifluoro-1-(4-fluoro-3-methylphenyl)ethanone

63297-1G 63297-25G 63297-5G 63297-500MG
76892-754EA 84.88 USD
76892-754 76892-756 76892-758 77647-842
3'-Methyl-2,2,2,4'-tetrafluoroacetophenone ≥97%
3'-Methyl-2,2,2,4'-tetrafluoroacetophenone

Precaución: for Research Use Only

Formula: C₉H₆F₄O
Storage Temperature: Ambient
MDL Number: MFCD01320004
CAS Number: 845823-10-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR