3-Iodo-2-methylbenzotrifluoride ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 1-Iodo-2-methyl-3-(trifluoromethyl)benzene

62363-1G 62363-25G 62363-5G 62363-500MG
76892-588EA 94.43 USD
76892-588 76892-590 76892-592 77647-828
3-Iodo-2-methylbenzotrifluoride ≥97%
3-Iodo-2-methylbenzotrifluoride

Precaución: for Research Use Only

Formula: C₈H₆F₃I
Storage Temperature: Ambient
MDL Number: MFCD09800791
CAS Number: 863111-55-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR