3'-Bromo-5'-methylacetophenone ≥97%

Supplier: AOBChem USA

Warning

62333-1G 62333-25G 62333-500MG 62333-5G
76892-574EA 144.29 USD
76892-574 76892-576 76892-578 76892-580
3'-Bromo-5'-methylacetophenone ≥97%
3'-Bromo-5'-methylacetophenone

Precaución: for Research Use Only

Formula: C₉H₉BrO
Storage Temperature: Ambient
MDL Number: MFCD18905445
CAS Number: 1379325-64-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR