(2-(Dimethylamino)pyridin-4-yl)boronic acid ≥97%
Supplier: AOBChem USA
|
Warning
|
62129-1G
62129-500MG
62129-5G
62129-25G
76892-560EA
563.34
USD
76892-560
76892-562
76892-564
77656-706
(2-(Dimethylamino)pyridin-4-yl)boronic acid ≥97%
(2-(Dimethylamino)pyridin-4-yl)boronic acid
Formula:
C₇H₁₁BN₂O₂ MW: 165.9854 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD13188980 CAS Number: 1207749-70-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...