2-(Benzyloxy)-3-methoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

61016-25G 61016-5G
76892-432EA 595.17 USD
76892-432 76892-434
2-(Benzyloxy)-3-methoxybenzaldehyde ≥97%
2-(Benzyloxy)-3-methoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₁₅H₁₄O₃
Storage Temperature: Ambient
MDL Number: MFCD00202674
CAS Number: 2011-06-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR