1-Fluoro-3-Methoxy-2-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

54587-1G 54587-25G 54587-500MG 54587-5G
76891-628EA 200.52 USD
76891-628 76891-630 76891-632 76891-634
1-Fluoro-3-Methoxy-2-methylbenzene ≥97%
1-Fluoro-3-Methoxy-2-methylbenzene
Formula: C₈H₉FO
MW: 140.15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12922638
CAS Number: 1159883-21-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR