1,2-Dibromo-3,5-dichlorobenzene ≥97%

Supplier: AOBChem USA

Warning

49597-1G 49597-25G 49597-5G
76890-802EA 62.6 USD
76890-802 76890-804 76890-806
1,2-Dibromo-3,5-dichlorobenzene ≥97%
1,2-Dibromo-3,5-dichlorobenzene

Precaución: for Research Use Only

Formula: C₆H₂Br₂Cl₂
Storage Temperature: Ambient
CAS Number: 81067-40-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR