2-(N,N-DiBOC-Amino)-5-bromopyridine ≥97%

Supplier: AOBChem USA

Warning

48180-1G 48180-25G 48180-500MG 48180-5G
76890-414EA 132.62 USD
76890-414 76890-416 76890-418 76890-420
2-(N,N-DiBOC-Amino)-5-bromopyridine ≥97%
2-(N,N-DiBOC-Amino)-5-bromopyridine
Formula: C₁₅H₂₁BrN₂O₄
Storage Temperature: Refrigerator
MDL Number: MFCD18249958
CAS Number: 209959-28-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR