1,2-Dibromo-5-methyl-3-nitrobenzene ≥97%

Supplier: AOBChem USA

Warning

47768-1G 47768-25G 47768-500MG 47768-5G
76890-358EA 182.48 USD
76890-358 76890-360 76890-362 76890-364
1,2-Dibromo-5-methyl-3-nitrobenzene ≥97%
1,2-Dibromo-5-methyl-3-nitrobenzene
Formula: C₇H₅Br₂NO₂
Storage Temperature: Ambient
MDL Number: MFCD18089295
CAS Number: 73557-63-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR