≥97%

Supplier: AOBChem USA

Warning

35524-25G 35524-5G
76888-052EA 453.01 USD
76888-052 76888-054
≥97%
3,5-Dibromo-4-methylanisole

Precaución: for Research Use Only

Formula: C₈H₈Br₂O
MW: 279.96
Storage Temperature: Ambient
MDL Number: MFCD00013526
CAS Number: 14542-71-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR