2-Chloro-3,6-difluorobenzyl alcohol ≥97%

Supplier: AOBChem USA

Warning

34017-25G 34017-5G
76887-722EA 418 USD
76887-722 76887-724
2-Chloro-3,6-difluorobenzyl alcohol ≥97%
2-Chloro-3,6-difluorobenzyl alcohol

Precaución: for Research Use Only

Formula: C₇H₅ClF₂O
Storage Temperature: Ambient
MDL Number: MFCD01631445
CAS Number: 261762-44-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR