2-(Benzyloxy)-4-iodopyridine ≥97%

Supplier: AOBChem USA

Warning

31999-1G 31999-25G 31999-500MG 31999-5G
76887-366EA 379.81 USD
76887-366 76887-368 76887-370 76887-372
2-(Benzyloxy)-4-iodopyridine ≥97%
2-(Benzyloxy)-4-iodopyridine
Formula: C₁₂H₁₀INO
Storage Temperature: Refrigerator
MDL Number: MFCD26401038
CAS Number: 896155-81-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR