1,4-Di(4-pyridyl)benzene ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4,4'-(1,4-Phenylene)bispyridine, 1,4-Di(pyridin-4-yl)benzene

27065-1G 27065-25G 27065-500MG 27065-5G
76886-114EA 203.7 USD
76886-114 76886-116 76886-118 76886-120
1,4-Di(4-pyridyl)benzene ≥97%
1,4-Di(4-pyridyl)benzene

Precaución: for Research Use Only

Formula: C₁₆H₁₂N₂
MW: 232.29 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00445845
CAS Number: 113682-56-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR