1,6-Dibromo-2-methoxynaphthalene ≥97%

Supplier: AOBChem USA

Warning

24667-25G 24667-5G
76885-518EA 466.8 USD
76885-518 76885-520
1,6-Dibromo-2-methoxynaphthalene ≥97%
1,6-Dibromo-2-methoxynaphthalene
Formula: C₁₁H₈Br₂O
Storage Temperature: Ambient
MDL Number: MFCD00230831
CAS Number: 66996-59-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR