2-Iodo-6-nitrotoluene ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 1-Iodo-2-methyl-3-nitrobenzene

23190-25G 23190-5G
76885-162EA 580.32 USD
76885-162 76885-164
2-Iodo-6-nitrotoluene ≥97%
2-Iodo-6-nitrotoluene

Precaución: for Research Use Only

Formula: C₇H₆INO₂
MW: 263.300 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08272202
CAS Number: 41252-98-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR