1-Benzyloxy-3,5-difluorobenzene ≥97%

Supplier: AOBChem USA

Warning

15460-1G 15460-25G 15460-5G
76883-068EA 75.33 USD
76883-068 76883-070 76883-072
1-Benzyloxy-3,5-difluorobenzene ≥97%
1-Benzyloxy-3,5-difluorobenzene
Formula: C₁₃H₁₀F₂O
Storage Temperature: Refrigerator
MDL Number: MFCD18072457
CAS Number: 176175-97-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR