4-Methoxy-3-(methoxymethyl)phenylboronic acid ≥95% research grade

Supplier: AOBChem USA

Warning

31997-1G 31997-25G 31997-500MG 31997-5G
76930-774EA 379.81 USD
76930-774 76930-776 76930-778 76930-780
4-Methoxy-3-(methoxymethyl)phenylboronic acid ≥95% research grade
4-Methoxy-3-(methoxymethyl)phenylboronic acid
Formula: C₉H₁₃BO₄
MW: 196.008 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14687257
CAS Number: 338454-42-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR