5,7-Dibromo-1H-indazole ≥95%

Supplier: AOBChem USA

Warning

21012-1G 21012-25G 21012-500MG 21012-5G
76927-290EA 195.21 USD
76927-290 76927-292 76927-294 76927-296
5,7-Dibromo-1H-indazole ≥95%
5,7-Dibromo-1H-indazole
Formula: C₇H₄Br₂N₂
MW: 275.928 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD05664000
CAS Number: 50477-28-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR