1-Bromo-5-chloro-4-iodo-2-(trifluoromethyl)benzene ≥95% research grade

Supplier: AOBChem USA

Warning

Synonyms: 2-Bromo-4-chloro-5-iodobenzotrifluoride

24398-1G 24398-25G 24398-5G
76899-432EA 79.57 USD
76899-432 76899-434 76899-436
1-Bromo-5-chloro-4-iodo-2-(trifluoromethyl)benzene ≥95% research grade
1-Bromo-5-chloro-4-iodo-2-(trifluoromethyl)benzene
Formula: C₇H₂BrClF₃I
Storage Temperature: Refrigerator
MDL Number: MFCD22543694
CAS Number: 1394291-54-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR