4-Bromo-2-chloropyridin-3-ol ≥95%

Supplier: AOBChem USA

Warning

26252-1G 26252-25G 26252-500MG 26252-5G
76912-284EA 439.22 USD
76912-284 76912-286 76912-288 76912-290
4-Bromo-2-chloropyridin-3-ol ≥95%
4-Bromo-2-chloropyridin-3-ol
Formula: C₅H₃BrClNO
MW: 208.44 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD16606310
CAS Number: 1211583-55-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR