2,4,5-Trichlorotoluene ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 1,2,4-Trichloro-5-methylbenzene, 1-Methyl-2,4,5-trichlorobenzene

63063-1G 63063-25G 63063-500MG 63063-5G
76892-674EA 334.19 USD
76892-674 76892-676 76892-678 76892-680
2,4,5-Trichlorotoluene ≥95%
2,4,5-Trichlorotoluene

Precaución: for Research Use Only

Formula: C₇H₅Cl₃
MW: 195.47 g/mol
Storage Temperature: Ambient
CAS Number: 6639-30-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR