1,4-Dibromo-2-chloro-5-fluorobenzene ≥95%

Supplier: AOBChem USA

Warning

60699-25G 60699-5G
76892-380EA 595.17 USD
76892-380 76892-382
1,4-Dibromo-2-chloro-5-fluorobenzene ≥95%
1,4-Dibromo-2-chloro-5-fluorobenzene
Formula: C₆H₂Br₂ClF
MW: 288.339 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09878159
CAS Number: 1000572-83-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR