Sodium chloride 0.9% USP for injection
Supplier: BAXTER IV SOLUTIONS
Synonyms:
Halite
, Sea salt
, NaCl
, Saline
UE1323D
75840-946CS
3033.22
USD
75840-946
Sodium chloride 0.9% USP for injection
Sodium chloride
Sterile solutions for injection.
- Intravenous use
- Subcutaneous use
- mEq/L: Na 154, Cl 154
- mOsmol/L (calc): 308
- NDC Number: 0338-9543-04
- pH: 5.0 (4.5 – 7.0)
Envase: Multi Pack contains 16 bags per overwrap pouch. Quad Pack contains four bags per overwrap pouch. Single Pack contains one bag per overwrap pouch.
Formula:
NaCl MW: 58.44 g/mol |
MDL Number:
MFCD00003477 CAS Number: 7647-14-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...