3-Formylindole-5-boronic acid pinacol ester, N-BOC protected 98%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 3-formyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole-1-carboxylate

A997048-100MG A997048-1G A997048-250MG
76881-074EA 29 USD
76881-074 76881-076 76881-078
3-Formylindole-5-boronic acid pinacol ester, N-BOC protected 98%
3-Formylindole-5-boronic acid pinacol ester, N-BOC protected

Precaución: for Research Use Only

Formula: C₂₀H₂₆BNO₅
MDL Number: MFCD07364063
CAS Number: 1025707-92-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR