rel-(1R,2r,3S,4R,5s,6S)-Cyclohexane-1,2,3,4,5,6-hexayl hexanicotinate 97%

Supplier: Ambeed
Warning

Synonyms: Inositol nicotinate

A991083-100G A991083-25G A991083-25MG A991083-10MG
76881-060EA 42.44 USD
76881-060 76881-062 76881-064 77614-714
rel-(1R,2r,3S,4R,5s,6S)-Cyclohexane-1,2,3,4,5,6-hexayl hexanicotinate 97%
rel-(1R,2r,3S,4R,5s,6S)-Cyclohexane-1,2,3,4,5,6-hexayl hexanicotinate

Precaución: for Research Use Only

Formula: C₄₂H₃₀N₆O₁₂
MW: 810.72 g/mol
MDL Number: MFCD00006387
CAS Number: 6556-11-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR