3-(Cyclopropylmethoxy)-5-methylphenylboronic acid pinacol ester 96%

Supplier: Ambeed
Warning

Synonyms: 2-(3-(Cyclopropylmethoxy)-5-methylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

A990114-100MG A990114-1G A990114-250MG
76881-054EA 37.14 USD
76881-054 76881-056 76881-058
3-(Cyclopropylmethoxy)-5-methylphenylboronic acid pinacol ester 96%
3-(Cyclopropylmethoxy)-5-methylphenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₇H₂₅BO₃
MW: 288.1896 g/mol
MDL Number: MFCD12546604
CAS Number: 1218789-80-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR