tert-Butyl 8-oxa-2,5-diazaspiro[3.5]nonane-2-carboxylate ≥97%

Supplier: Ambeed
Warning

A979666-100MG A979666-1G
76881-038EA 189 USD
76881-038 76881-040
tert-Butyl 8-oxa-2,5-diazaspiro[3.5]nonane-2-carboxylate ≥97%
tert-Butyl 8-oxa-2,5-diazaspiro[3.5]nonane-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₂₀N₂O₃
MDL Number: MFCD14581204
CAS Number: 1251002-01-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR