Piperidine-2-carbonitrile hydrochloride 95%

Supplier: Ambeed
Warning

A979313-100MG A979313-1G A979313-250MG
76881-032EA 45.62 USD
76881-032 76881-034 76881-036
Piperidine-2-carbonitrile hydrochloride 95%
Piperidine-2-carbonitrile hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₁ClN₂
MW: 146.61 g/mol
MDL Number: MFCD11840203
CAS Number: 117921-54-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR