2-Bromo-1-(tetrahydro-2H-pyran-4-yl)ethanone 95%

Supplier: Ambeed
Danger

A970168-10G A970168-1G A970168-250MG A970168-5G
76881-022EA 941.02 USD
76881-022 76881-024 76881-026 76881-028
2-Bromo-1-(tetrahydro-2H-pyran-4-yl)ethanone 95%
2-Bromo-1-(tetrahydro-2H-pyran-4-yl)ethanone

Precaución: for Research Use Only

Formula: C₇H₁₁BrO₂
MW: 207.065 g/mol
MDL Number: MFCD11656452
CAS Number: 141095-78-5
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR