Ethyl-7-bromoimidazo[1,2-a]pyridine-2-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: Ethyl 7-bromoimidazo[1,2-a]pyridine-2-carboxylate

A968233-250MG A968233-5G A968233-1G A968233-100MG
76881-016EA 30.77 USD
76881-016 76881-018 77595-558 77674-424
Ethyl-7-bromoimidazo[1,2-a]pyridine-2-carboxylate 98%
Ethyl 7-bromoimidazo[1,2-a]pyridine-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₉BrN₂O₂
MW: 269.10 g/mol
MDL Number: MFCD11505036
CAS Number: 1187236-18-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR