2-Amino-5-nitropyrimidine-4,6-diol 95%

Supplier: Ambeed
Warning

Synonyms: 2-Amino-4,6-dihydroxy-5-nitro-1,3-diazine, 2-Amino-4,6-dihydroxy-5-nitropyrimidine

A967565-100MG A967565-250MG
76881-012EA 5.31 USD
76881-012 76881-014
2-Amino-5-nitropyrimidine-4,6-diol 95%
2-Amino-5-nitropyrimidine-4,6-diol

Precaución: for Research Use Only

Formula: C₄H₄N₄O₄
MW: 172.099 g/mol
MDL Number: MFCD00082801
CAS Number: 80466-56-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR