Methyl-1-(4-bromophenyl)cyclopropanecarboxylate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 1-(4-bromophenyl)cyclopropanecarboxylate

A958126-1G A958126-250MG A958126-5G
76880-978EA 102.91 USD
76880-978 76880-980 76880-982
Methyl-1-(4-bromophenyl)cyclopropanecarboxylate 98%
Methyl 1-(4-bromophenyl)cyclopropanecarboxylate

Precaución: for Research Use Only

Formula: C₁₁H₁₁BrO₂
MW: 255.11 g/mol
MDL Number: MFCD12913964
CAS Number: 638220-35-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR