2,6-Dimethyl-3-nitro-4-pyridinol 98%

Supplier: Ambeed
Warning

Synonyms: 2,6-Dimethyl-3-nitropyridin-4-ol

A955132-1G A955132-5G A955132-250MG
76880-958EA 26 USD
76880-958 76880-960 77884-892
2,6-Dimethyl-3-nitro-4-pyridinol 98%
2,6-Dimethyl-3-nitro-4-pyridinol

Precaución: for Research Use Only

Formula: C₇H₈N₂O₃
MDL Number: MFCD18253578
CAS Number: 13603-45-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR