2-Fluoro-4-hydroxybenzeneboronic acid pinacol ester 98%

Supplier: Ambeed
Warning

A942858-100MG A942858-1G A942858-250MG A942858-5G
76880-918EA 29.71 USD
76880-918 76880-920 76880-922 76880-924
2-Fluoro-4-hydroxybenzeneboronic acid pinacol ester 98%
2-Fluoro-4-hydroxybenzeneboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₂H₁₆BFO₃
MW: 238.065 g/mol
MDL Number: MFCD16994421
CAS Number: 1029439-02-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR