3-(Cyclohexylcarbamoyl)-4-fluorophenylboronic acid 98%

Supplier: Ambeed
Danger

A939726-1G A939726-250MG A939726-5G
76880-912EA 63 USD
76880-912 76880-914 76880-916
3-(Cyclohexylcarbamoyl)-4-fluorophenylboronic acid 98%
3-(Cyclohexylcarbamoyl)-4-fluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₇BFNO₃
MDL Number: MFCD08235043
CAS Number: 874219-24-6
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR