Methyl 2-bromo-3,4-methylenedioxy-5-methoxybenzoate 97%

Supplier: Ambeed
Warning

A939620-100MG A939620-1G A939620-250MG
76880-906EA 44 USD
76880-906 76880-908 76880-910
Methyl 2-bromo-3,4-methylenedioxy-5-methoxybenzoate 97%
Methyl 2-bromo-3,4-methylenedioxy-5-methoxybenzoate

Precaución: for Research Use Only

Formula: C₁₀H₉BrO₅
MDL Number: MFCD00792457
CAS Number: 81474-46-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR