(1R,2S)-2-(tert-Butoxycarbonylamino)cyclopentanecarboxylic acid 98%

Supplier: Ambeed
Warning

Synonyms: (1R,2S)-2-((tert-Butoxycarbonyl)amino)cyclopentanecarboxylic acid

A930077-100MG A930077-250MG
76880-896EA 103 USD
76880-896 76880-898
(1R,2S)-2-(tert-Butoxycarbonylamino)cyclopentanecarboxylic acid 98%
(1R,2S)-2-(tert-Butoxycarbonylamino)cyclopentanecarboxylic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₉NO₄
MDL Number: MFCD09750530
CAS Number: 130981-12-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR