1,3-Dibromo-5,5-dimethylhydantoin 97%

Supplier: Ambeed
Danger

A929052-25G A929052-100G
76880-894EA 16 USD
76880-894 77614-648
1,3-Dibromo-5,5-dimethylhydantoin 97%
1,3-Dibromo-5,5-dimethylhydantoin

Precaución: for Research Use Only

Formula: C₅H₆Br₂N₂O₂
MDL Number: MFCD00003189
CAS Number: 77-48-5
UN: 3087
ADR: 5.1,6.1,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR